missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ NADP, Monosodium Salt

Catalog No. 4819711GM
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4819711GM 1 g
481971500MG 500 mg
4819715GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 4819711GM Supplier MilliporeSigma™ Supplier No. 4819711GM
Only null left

Participates in oxidation-reduction reactions

  • Coenzyme that is widely distributed in living matter

Specifications

Content And Storage -20°C
Additive NADP, Monosodium Salt
Quantity 1 g