missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Nω-Hydroxy-nor-L-arginine, Diacetate Salt

Catalog No. 3992751mg
Change view
Click to view available options
Quantity:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3992751MG 1mg
3992755MG 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3992751MG Supplier MilliporeSigma™ Supplier No. 3992751MG
Only null left

Serves as potent, selective, competitive, high affinity inhibitor of arginase.

EMD Millipore Calbiochem™ Nω-Hydroxy-nor-L-arginine, Diacetate Salt is shown to specifically interact with manganese-cluster of enzyme active site.

  • Inhibits arginase from rat liver (IC50 = 2μM) mouse macrophages (IC50 = 50μM)
  • Does not function as substrate or inhibitor for NOS
  • Useful tool to study interaction between arginase NOS
  • CAS: 189302-40-7
  • Molecular formula: C5H12N4O3 · 2CH3CO2H
  • Color: Off-White

Specifications

For Use With (Application) Arginase from rat liver
Content And Storage −20°C
Product Type Nω-Hydroxy-nor-L-arginine, Diacetate Salt, Packaged in Glass Bottle
Quantity 1mg