missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ MOWIOL™ 4-88 Reagent

Catalog No. 475904100GM
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
475904100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 475904100GM Supplier MilliporeSigma™ Supplier No. 475904100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for mounting cells on slides for detection of cell-surface antigens.

EMD Millipore Calbiochem™ MOWIOL™ 4-88 Reagent is used as an anti-fading agent.

  • Detects by immunofluorescence
  • Used as anti-fading agent
  • CAS: 9002-89-5
  • Packaging: Glass Bottle
  • Color: Yellow

Specifications

Description MOWIOL 4-88 Reagent
Quantity 100 g
Content And Storage 15°C to 30°C