accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
Sign In
Don't have an account?
Create an Account
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Microbiological Media and Media Additives
Media Additives
42758450GM
Print
MilliporeSigma™ Calbiochem™ Lactulose
Catalog No.
42758450GM
Change view
Chip view icon
List view icon
Click to view available options
Lactulose; Qty: 50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
42758450GM
Lactulose; Qty: 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No.
42758450GM
Supplier
MilliporeSigma™
Supplier No.
42758450GM
Sign In
or
Register
to check your price and availability.
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Description
Acts as β-galactosidase substrate
Specifications
Specifications
Content And Storage
15°C to 30°C
Additive
Lactulose
Quantity
50 g
Product Suggestions
Documents