missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ ImmunoPen™

Catalog No. 4021761EA
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4021761EA 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4021761EA Supplier MilliporeSigma™ Supplier No. 4021761EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Useful in immunostaining techniques on paraffin, frozen sections cytology preparations.

EMD Millipore Calbiochem™ ImmunoPen™ forms hydrophobic barrier retains antibody.

  • Sufficient for 500 to 800 individual applications
  • Packaging: Glass Bottle

Specifications

Description ImmunoPen
Quantity 1 Ea.
Content And Storage 15°C to 30°C