missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Human Plasma Fibronectin

Catalog No. 3416355MG
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3416355MG 5 mg
3416351MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3416355MG Supplier MilliporeSigma™ Supplier No. 3416355MG

Promotes attachment of cells to commonly used culture substrates.

EMD Millipore Calbiochem™ Human Plasma Fibronectin is purified from human plasma.

  • Native fibronectin
  • Contaminants: None
  • Detected by IEP with antiserum to fibronectin or normal human serum
  • Single Major B by SDS-PAGE
  • CAS: 86088-83-7

Specifications

Content And Storage -20°C
Product Type Human Plasma Fibronectin
Formulation In PBS, pH 7.3.
Quantity 5 mg