missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Human Galectin-3C, Recombinant, E.coli
Description
- Expressed in E. coli and subsequently cleaved with Clostridium histolyticum collagenase type VII to yield Galectin-3C
- Purified by affinity chromatography
Specifications
Specifications
| Formulation | Lyophilized from deionized water, 2μg lactose/μg lectin. |
| Quantity | 100 μg |
| Storage Requirements | -20°C |
| Endotoxin Concentration | ≤1 EU/μg protein |
| Product Type | Human Galectin-3C, Recombinant, E.coli |
| Form | Lyophilized |
| Purity or Quality Grade | ≥95% by SDS-PAGE |
For Research Use Only