missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ High Purity IPTG, Animal-Free
Description
- Contains no animal-derived products
- Inducer of β-D-galactosidase
Specifications
Specifications
| Physical Form | White to off-white solid |
| Type | IPTG |
| Quantity | 5 g |
For Research Use Only