Learn More
MilliporeSigma™ Calbiochem™ EDAC, Hydrochloride
Used for conjugating haptens to proteins polypeptides.
$178.28 - $501.74
Specifications
Content And Storage −20°C
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
34100625GM
View Documents Promotion Details
MilliporeSigma™
34100625GM
25gm
Each for $501.74
Only null left
3410065GM
View Documents Promotion Details
MilliporeSigma™
3410065GM
5mg
Each for $178.28
Only null left
Description
Description
EMD Millipore Calbiochem™ EDAC, Hydrochloride is water-soluble derivative of carbodiimide.
  • Modifies NMDA receptors
  • Condensing agent in peptide synthesis
  • Water soluble carbodiimide derivative
Specifications
Specifications
−20°C