missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ EDAC, Hydrochloride

Catalog No. 3410065GM
Change view
Click to view available options
Quantity:
25gm
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3410065GM 5mg
34100625GM 25gm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3410065GM Supplier MilliporeSigma™ Supplier No. 3410065GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for conjugating haptens to proteins polypeptides.

EMD Millipore Calbiochem™ EDAC, Hydrochloride is water-soluble derivative of carbodiimide.

  • Modifies NMDA receptors
  • Condensing agent in peptide synthesis
  • Water soluble carbodiimide derivative
  • CAS: 25952-53-8
  • Molecular formula: C8H17N3 · HCl

Specifications

Quantity 5mg
Content And Storage −20°C