missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ EDAC, Hydrochloride

Used for conjugating haptens to proteins polypeptides.

Supplier:  MilliporeSigma™ 34100625GM

 View more versions of this product

Catalog No. 34100625GM

Only null left
Explore available promotions

Description

Description

EMD Millipore Calbiochem™ EDAC, Hydrochloride is water-soluble derivative of carbodiimide.

  • Modifies NMDA receptors
  • Condensing agent in peptide synthesis
  • Water soluble carbodiimide derivative
  • CAS: 25952-53-8
  • Molecular formula: C8H17N3 · HCl
Specifications

Specifications

25gm
−20°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions