missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ DM-NITROPHEN™ Reagent, Tetrasodium Salt
Description
- Can be microinjected or used with permeabilized cells
- May also be useful as caged Mg2+ chelator. Kd (Ca2+) = 5nm; Kd (Mg2+) = 2-5μM
Specifications
Specifications
| Content And Storage | -20°C |
| Form | Solid |
| Product Type | Calbiochem DM-NITROPHEN Reagent |
| Quantity | 5 mg |
| Purity | ≥93% (HPLC) |
| Solubility | Water |
| Synonym | Dimethoxynitrophenamine, 4Na, 1-(2-Nitro-4,5-dimethoxyphenyl)-1,2-diaminoethane-N,N,Nʹ,Nʹ-tetraacetic Acid, 4Na |