missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Dextran Sulfate, Sodium Salt, Molecular Biology Grade

Catalog No. 26515210GM
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26515210GM 10 g
26515250GM 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26515210GM Supplier MilliporeSigma™ Supplier No. 26515210GM

Used as anticoagulant.

EMD Millipore Calbiochem™ Dextran Sulfate, Sodium Salt, Molecular Biology Grade is long chain polymer of sulfated glucose.

  • Contains 17 to 20% sulfur
  • Inhibits binding of HIV-1 to CD4+ T lymphocytes
  • Accelerates hybridization of DNA probes to immobilized nucleic acids
  • Contaminants DNases, proteases, RNases: None Detected
  • CAS: 9011-18-1
  • Molecular Biology Grade
  • Color: Off-White

Specifications

Content And Storage 2°C to 8°C
Description Dextran Sulfate, Sodium Salt; Solid
Quantity 10 g