missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ DAPI Stain

Catalog No. 5087410001
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5087410001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5087410001 Supplier MilliporeSigma™ Supplier No. 5.08741.0001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Serves as sterile-filtered solution of DAPI in water.

  • Cells are typically stained at 1μg/mL
  • Cell permeable, reversible
  • Assay: ≥97% by HPLC
  • CAS: 28718-90-3
  • Molecular Formula: C16H15N5 · 2HCl
  • Packaging: Glass Bottle

Specifications

Product Type DAPI Stain
Quantity 5 mg
For Use With (Application) Flow Cytometry
Content And Storage -20°C