missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ β-Cyclodextrin
Description
Homogeneous cyclic α1,4-linked D-glucopyranose units in a seven-member ring. Forms clathrates. Suitable for use with dansyl chloride to form water-soluble complexes for fluorescent labeling of proteins.
Specifications
Specifications
| Quantity | 500 g |
| Molecular Formula | C42H70O35 |
| Content And Storage | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C |
| Format | Solid |
| Color | White |