missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ β-Cyclodextrin

Catalog No. 239760500GM
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
239760500GM 500 g
23976025GM 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 239760500GM Supplier MilliporeSigma™ Supplier No. 239760500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Homogeneous cyclic α1,4-linked D-glucopyranose units in a seven-member ring. Forms clathrates. Suitable for use with dansyl chloride to form water-soluble complexes for fluorescent labeling of proteins.

Homogeneous cyclic α1,4-linked D-glucopyranose units in a seven-member ring. Forms clathrates. Suitable for use with dansyl chloride to form water-soluble complexes for fluorescent labeling of proteins.

Specifications

Quantity 500 g
Molecular Formula C42H70O35
Content And Storage Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C
Format Solid
Color White