missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Cholesterol/Cholesteryl Ester Quantitation Kit

Catalog No. 4289011KIT
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4289011KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4289011KIT Supplier MilliporeSigma™ Supplier No. 4289011KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Cholesterol Reaction buffer, Cholesterol Probe, Dimethyl sulfoxide, Enzyme Mix, Cholesterol Esterase, Cholesterol standard and user protocol

Measures cholesterol cholesteryl ester quantitatively.

EMD Millipore Calbiochem™ Cholesterol/Cholesteryl Ester Quantitation Kit is sensitive assay.

  • Detects by colorimetric or fluorometric method
  • Fluorometric assay can detect 0.02-1μg/well
  • Packaging: Glass Bottle

Specifications

Assay Range 0.2 to 10μg/well
Includes Cholesterol Reaction Buffer, Cholesterol Probe, Dimethyl sulfoxide, Enzyme Mix, Cholesterol Esterase, Cholesterol Standard, and user protocol
Quantity 1 Kit
Sample Type Cells, tissues, serum
Content And Storage -20°C
Product Type Cholesterol/Cholesteryl Ester Quantitation Kit
No. of Assays 100 Tests