missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ CB-Protein Assay™ Kit

Catalog No. 2194681KIT
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2194681KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2194681KIT Supplier MilliporeSigma™ Supplier No. 2194681KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

CB-Protein Dye, Albumin standard and user protocol

Used for estimation of protein concentration.

EMD Millipore Calbiochem™ CB-Protein Assay™ Kit is compatible with reducing reagents.

  • Not suitable for use with solutions containing detergents
  • Based on improved Coomassie dye reagent
  • Packaging: Glass Bottle

Specifications

Product Type CB-Protein Assay Kit
Sufficient For 500 tube assays or 5000 microwell plate assays
Kit Contents Albumin standard, CB-Protein dye, User protocol
Quantity 1 Kit
Storage Requirements 2°C to 8°C