missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Calpain Activity Assay Kit, Fluorogenic

Catalog No. QIA1201KIT
Change view
Click to view available options
Quantity:
96 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QIA1201KIT 96 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QIA1201KIT Supplier MilliporeSigma™ Supplier No. QIA1201KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Actively drive reagents through membrane.

  • Vacuum-driven technology
  • Detection method: Chemiluminescence Fluorescence
  • Built-in flow distributor
  • Actively drive reagents through membrane
  • Packaging: Glass Bottle

Specifications

Cross Reactivity Mammals
Quantity 96 Tests/Kit
Form Liquid