Learn More
MilliporeSigma™ Calbiochem™ Albumin, Human Serum, Fraction V, Low Heavy Metals
Prepared under non-denaturing conditions by modification of Cohn procedure
Supplier: MilliporeSigma™ 12666100GM
Specifications
| Protein Standard, Protein Base or Filler, Stabilizer and Diluent in Serology and Immunohematology | |
| Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV. | |
| Solid |
| 100 g | |
| 2° to 8°C | |
| ≥95% by SDS PAGE |
For Research Use Only