missing translation for 'onlineSavingsMsg'
Learn More

Caffeine 98.0+%, TCI America™

Catalog No. p-7125251
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C204225G 25 g
C2042500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C204225G Supplier TCI America Supplier No. C204225G
Only null left

Chemical Identifiers

CAS 58-08-2
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.19
MDL Number MFCD00005758
InChI Key RYYVLZVUVIJVGH-UHFFFAOYSA-N
Synonym caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
PubChem CID 2519
ChEBI CHEBI:27732
IUPAC Name 1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
SMILES CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Melting Point 238°C
Color White
pH 6.9 (1% H2O soln.)
Quantity 25 g
UN Number 1544
Formula Weight 194.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Caffeine
EINECSNumber (9)-0419
RTECSNumber EV6475000
TSCA Yes