missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caffeine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 58-08-2 |
| Physical Form | Neat |
| Linear Formula | C8H10N4O2 |
| MDL Number | MFCD00005758 |
| Melting Point | 234°C to 236.5°C |
| Synonym | 1,3,7-Trimethylxanthine |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 194.19 |
| Grade | Certified Reference Material |