missing translation for 'onlineSavingsMsg'
Learn More

Caffeine Impurity F Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014881
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014881 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014881 Supplier Merck Emd Millipore Supplier No. PHR200050MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Pharmaceutical (small molecule)

Specifications

CAS 611-59-6
Linear Formula C7H8N4O2
MDL Number MFCD00005727
Melting Point 294°C to 296°C
Synonym 1,7-Dimethylxanthine; 1,7-DIMETHYL-3,7-DIHYDRO- 1 H-PURINE-2,6-DIONE; 1,7-Dimethyl-1 H-purine-2,6-dione; 1,7-Dimethyl-3,7-dihydro-1 H-purine-2,6-dione; 2,6-Dihydroxy-1,7-dimethylpurine; NSC 400018; Paraxanthine
Type Certified Reference Material
Quantity 50 mg
Formula Weight 180.16
Grade Certified Reference Material