missing translation for 'onlineSavingsMsg'
Learn More

Caffeine, Anhydrous, USP, Spectrum™ Chemical

Catalog No. 18621730
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18621730 50 kg
18621726 125 g
18621727 500 g
18621728 2.5 kg
18621729 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18621730 Supplier Spectrum Chemical Mfg Cor Supplier No. C403650KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58-08-2

Specifications

Packaging Drum
Quantity 50 kg
Molecular Weight (g/mol) 194.19
Percent Purity 98.5%
Grade USP
Physical Form Crystals or Flakes
Chemical Name or Material Caffeine, Anhydrous