Learn More
Caffeine, 99.7%
CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol
Supplier: Thermo Scientific Chemicals 03921422
Description
It acts as a stimulant drug and a reversible acetylcholinesterase inhibitor. It plays an important role for the inactivation of Escherichia coli O 157:H 7 in brain heart infusion (BHI) broth. It is also useful for the protection from photodamage after ultraviolet irradiation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Caffeine | |
| 234°C to 236°C | |
| 178°C (sublimation) | |
| C8H10N4O2 | |
| 100 g | |
| caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep | |
| RYYVLZVUVIJVGH-UHFFFAOYSA-N | |
| 1,3,7-trimethylpurine-2,6-dione | |
| 2519 | |
| 194.19 |
| 58-08-2 | |
| 1.23 g/mL | |
| 99.7% | |
| MFCD00005758 | |
| 17705 | |
| Soluble in water,pyridine and ethyl acetate. Partially soluble in acetone,alcohol and petroleum ether. | |
| CN1C=NC2=C1C(=O)N(C)C(=O)N2C | |
| 194.19 | |
| CHEBI:27732 | |
| Powder |
Chemical Identifiers
| 58-08-2 | |
| 194.19 | |
| RYYVLZVUVIJVGH-UHFFFAOYSA-N | |
| 2519 | |
| 1,3,7-trimethylpurine-2,6-dione |
| C8H10N4O2 | |
| MFCD00005758 | |
| caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep | |
| CHEBI:27732 | |
| CN1C=NC2=C1C(=O)N(C)C(=O)N2C |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 200-362-1
RTECSNumber : EV6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only