missing translation for 'onlineSavingsMsg'
Learn More

Caffeine, 98.5%, specified according to the req. of USP/BP

The crystalline powder form of the stimulant caffeine, CAS # 58-08-2, purified for analytical chemistry applications such as HPLC. | CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol

$86.06 - $86.06

Chemical Identifiers

CAS 58-08-2
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.19
MDL Number MFCD00005758
InChI Key RYYVLZVUVIJVGH-UHFFFAOYSA-N
Synonym caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
PubChem CID 2519
ChEBI CHEBI:27732
IUPAC Name 1,3,7-trimethylpurine-2,6-dione
SMILES CN1C=NC2=C1C(=O)N(C)C(=O)N2C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC108160100
SDS View Documents
Thermo Scientific Chemicals
108160100
10 g Glass Bottle
Each for $86.06
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Caffeine is an alkaloid vagal/vasomotor stimulant found in certain nuts, seeds, and legumes
  • It is a methylxanthine, derived from a purine base compound found in the tissues of many organisms
  • Caffeine competitively binds adenosine receptors and prevents signaling from dampening, thereby sustaining activation, and producing increases in energy
Specifications

Chemical Identifiers

58-08-2
194.19
RYYVLZVUVIJVGH-UHFFFAOYSA-N
2519
1,3,7-trimethylpurine-2,6-dione
C8H10N4O2
MFCD00005758
caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
CHEBI:27732
CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

58-08-2
234.0°C to 239.0°C
1.2300g/mL
Authentic
Glass Bottle
MFCD00005758
26,461
15,1639
178 °C
meets the requirements
CN1C=NC2=C1C(=O)N(C)C(=O)N2C
194.19
CHEBI:27732
98.5%
Crystalline Powder or Crystals
0.2mL of 0.01M NaOH max.
White
0.5% max.
0.1% max. (total impurities) (HPLC)
C8H10N4O2
10 g
1.23
caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
Solubility in water: 20g/l (20°C). Other solubilities: 1g/66 mL alcohol,1g/50 mL acetone,freely soluble in pyrrole,soluble in ethyl acetate
RYYVLZVUVIJVGH-UHFFFAOYSA-N
1,3,7-trimethylpurine-2,6-dione
2519
194.19
0.5% max. (TLC)
Caffeine, 98.5%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-362-1

RTECSNumber : EV6475000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only