missing translation for 'onlineSavingsMsg'
Learn More

Caffeine, 1mg/ml in methanol

Catalog No. AC393540010
Change view
Click to view available options
Quantity:
1 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393540010 1 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC393540010 Supplier Thermo Scientific Chemicals Supplier No. 393540010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58-08-2
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.19
MDL Number MFCD00005758
InChI Key RYYVLZVUVIJVGH-UHFFFAOYSA-N
Synonym caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
PubChem CID 2519
ChEBI CHEBI:27732
IUPAC Name 1,3,7-trimethylpurine-2,6-dione
SMILES CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Density 0.7910g/mL
Flash Point 12°C
Packaging Glass bottle
Quantity 1 mL
Specific Gravity 0.791
Merck Index 15, 1639
Formula Weight 194.19
Chemical Name or Material Caffeine, 1mg/mL in methanol
Hazard Category GHS Signal Word: Danger
EINECSNumber 200-362-1

RUO – Research Use Only