missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Sulfate, Anhydrous, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7165271
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609426 25 g Amber Glass Bottle
18609423 125 g Amber Glass Bottle
18609425 500 g Amber Glass Bottle
18609424 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609426 Supplier Spectrum Chemical Mfg Cor Supplier No. C104725GM
Only null left

Chemical Identifiers

CAS 10124-36-4
Molecular Formula CdO4S
Molecular Weight (g/mol) 208.47
InChI Key QCUOBSQYDGUHHT-UHFFFAOYSA-L
IUPAC Name cadmium(2+) sulfate
SMILES [Cd++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade ACS
Loss on Drying 0.01
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.