accessibility menu, dialog, popup

UserName

CdOSOOO

CAS RN 10124-36-4

IUPAC Name: cadmium(2+) sulfate
Synonyms: cadmium sulfate kadmiumsulfat cadmiumsulfat sulfate de cadmium CdSO4 sulfato de cadmio
Molecular Weight (g/mol): 208.47
Molecular Formula: CdO4S
InChi Key: QCUOBSQYDGUHHT-UHFFFAOYSA-L
SMILES: [Cd++].[O-]S([O-])(=O)=O
Grade 
  • (2)
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (6)
Physical Form 
  • (1)
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (6)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)