missing translation for 'onlineSavingsMsg'
Learn More

Cadmium standard solution, for AAS, 1 mg/ml Cd in 2 to 5% HNO3

Catalog No. p-220981
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC195871000 100 mL
AC195875000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC195871000 Supplier Thermo Scientific Chemicals Supplier No. 195871000
Only null left

CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy

Specifications

Chemical Name or Material Cadmium standard solution
Packaging Plastic bottle
Name Note 1 mg/ml Cd in 0.5N HNO3
CAS 10022-68-1, 7697-37-2, 7732-18-5
Density 1.0120g/mL
Specific Gravity 1.012
Physical Form Solution
Molecular Formula CdH2N2O6
Concentration 997 to 1003 μg/mL of Cd(2+) in 0.5N HNO3 (20°C)
MDL Number MFCD00149626
Merck Index 15, 1627
Solubility Soluble in water
InChI Key IZIPRBSCGMISKX-UHFFFAOYSA-N
SMILES [Cd++].O[N+]([O-])=O.O[N+]([O-])=O
IUPAC Name cadmium(2+) bis(nitric acid)
Molecular Weight (g/mol) 238.44
Quantity 100 mL
PubChem CID 56924536
ChEBI CHEBI:86156
Formula Weight 308.48
Grade AAS
Show More Show Less

RUO – Research Use Only