missing translation for 'onlineSavingsMsg'
Learn More

Cadmium shot, 1-3mm (0.04-0.1in), Puratronic™, 99.9997% (metals basis)

Catalog No. p-4407303
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1135118 50 g
AA1135130 250 g
AA11351A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1135118 Supplier Thermo Scientific Chemicals Supplier No. 1135118
Only null left

CAS: 7440-43-9 | Cd | 112.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
SMILES [Cd]

Specifications

Color Silver-White
Appearance Silvery-white shot
Density 8.65g/cm3
Quantity 50 g
Vapor Pressure 0.00013 hPa
Form Shot
Assay Percent Range 99.9997% (Metals basis)
IUPAC Name cadmium
Chemical Name or Material Cadmium
Hazard Category H330-H341-H350-H361fd-H372-H500
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c
EINECSNumber 231-152-8
RTECSNumber EU9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only