Learn More
Cadmium selenide, 99.995% (metals basis), Thermo Scientific Chemicals
Used as a n-type semiconductor and its nanoparticles are used in laser diodes, opto-electronic devices, nanosensing, biomedical imaging and high efficiency solar cells. Since, it is transparent to infrared light, it finds application in photoresistors and windows for instruments utilizing IR light.
$1036.23 - $1036.23
Chemical Identifiers
| CAS | 1306-24-7 |
|---|---|
| Molecular Formula | CdSe |
| Molecular Weight (g/mol) | 191.39 |
| MDL Number | MFCD00010917 |
| InChI Key | AQCDIIAORKRFCD-UHFFFAOYSA-N |
| Synonym | cadmium selenide, cadmium ii selenide, cadmium 2+ selenide, cadmium selenide nanotubes, cadmium selenide trace metals basis 1g, cadmium selenide candot quantum dot core , 50umol/l in hexane, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene |
| PubChem CID | 14784 |
| ChEBI | CHEBI:50834 |
| IUPAC Name | selanylidenecadmium |
| SMILES | [Se]=[Cd] |
Description
Cadmium selenide is used as a n-type semiconductor and its nanoparticles are used in laser diodes, opto-electronic devices, nanosensing, biomedical imaging and high efficiency solar cells. Since, it is transparent to infrared light, it finds application in photoresistors and windows for instruments utilizing IR light.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1306-24-7 | |
| 191.39 | |
| AQCDIIAORKRFCD-UHFFFAOYSA-N | |
| 14784 | |
| selanylidenecadmium |
| CdSe | |
| MFCD00010917 | |
| cadmium selenide, cadmium ii selenide, cadmium 2+ selenide, cadmium selenide nanotubes, cadmium selenide trace metals basis 1g, cadmium selenide candot quantum dot core , 50umol/l in hexane, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene | |
| CHEBI:50834 | |
| [Se]=[Cd] |
Specifications
| >1350°C | |
| Powder | |
| CdSe | |
| UN3077 | |
| Insoluble in water. | |
| [Se]=[Cd] | |
| 191.39 | |
| CHEBI:50834 | |
| 99.995% | |
| 5.81 | |
| Cadmium selenide |
| 1306-24-7 | |
| 50 g | |
| MFCD00010917 | |
| cadmium selenide, cadmium ii selenide, cadmium 2+ selenide, cadmium selenide nanotubes, cadmium selenide trace metals basis 1g, cadmium selenide candot quantum dot core , 50umol/l in hexane, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene | |
| AQCDIIAORKRFCD-UHFFFAOYSA-N | |
| selanylidenecadmium | |
| 14784 | |
| 191.36 | |
| Plastic bottle | |
| 5.81 |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350-H373
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 215-148-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only