accessibility menu, dialog, popup

UserName

CdSe

CAS RN 1306-24-7

IUPAC Name: selanylidenecadmium
Synonyms: cadmium selenide
Molecular Weight (g/mol): 191.39
Molecular Formula: CdSe
InChi Key: AQCDIIAORKRFCD-UHFFFAOYSA-N
SMILES: [Se]=[Cd]
Percent Purity 
  • (2)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results