Learn More
Cadmium powder, -325 mesh, 99.5% (metals basis)
Cadmium powder, 99.5%, Cd, CAS Number-7440-43-9, ccris 112, unii-00bh33gngh, compounds, cadmium, elemental, cadmium and compounds, 00bh33gngh, kadmium, kadmium german, colloidal, hsdb 282, 500g, (metals basis), CHEBI:22977, Silver-White | CAS: 7440-43-9 | Cd | 112.41 g/mol
Supplier: Thermo Scientific Chemicals 01189122
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Silver-White | |
| 7440-43-9 | |
| −325 Mesh | |
| (metals basis) | |
| MFCD00010914 | |
| kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds | |
| [Cd] | |
| 112.41 | |
| CHEBI:22977 |
| Cadmium powder | |
| 100 g | |
| Solid | |
| Cd | |
| UN3179 | |
| BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
| cadmium | |
| 23973 | |
| 99.5% |
Chemical Identifiers
| 7440-43-9 | |
| 112.41 | |
| BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
| 23973 | |
| cadmium |
| Cd | |
| MFCD00010914 | |
| kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds | |
| CHEBI:22977 | |
| [Cd] |
Safety and Handling
GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
P201-P202-P210-P240-P241-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P370+P378q-P501c
H228-H330-H341-H350-H361fd-H372
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.
EINECSNumber : 231-152-8
RTECSNumber : EU9800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only