missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Iodide, Crystal, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C103512KG

 View more versions of this product

Catalog No. 18609412

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

7790-80-9
12 kg
OKIIEJOIXGHUKX-UHFFFAOYSA-L
cadmium(2+) diiodide
99%
Poly Pail
1
CdI2
[Cd++].[I-].[I-]
366.22
Reagent

Chemical Identifiers

7790-80-9
366.22
cadmium(2+) diiodide
CdI2
OKIIEJOIXGHUKX-UHFFFAOYSA-L
[Cd++].[I-].[I-]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.