missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Iodide, Crystal, 99%, Spectrum™ Chemical

Catalog No. p-7165270
Change view
Click to view available options
Quantity:
12 kg
2.5 kg
25 g
500 g
Packaging:
Amber Glass Bottle
Poly Pail
Catalog No. Quantity Packaging
18609415 25 g Amber Glass Bottle
18609413 500 g Amber Glass Bottle
18609414 2.5 kg Amber Glass Bottle
18609412 12 kg Poly Pail
4 options

Catalog No. 18609415

Supplier: Spectrum Chemical Mfg Cor C103525GM

Only null left

Chemical Identifiers

CAS 7790-80-9
Molecular Formula CdI2
Molecular Weight (g/mol) 366.22
InChI Key OKIIEJOIXGHUKX-UHFFFAOYSA-L
IUPAC Name cadmium(2+) diiodide
SMILES [Cd++].[I-].[I-]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.