missing translation for 'onlineSavingsMsg'
Learn More

Cadmium hydroxide, Puratronic™, 99.98% (metals basis)

CAS: 21041-95-2 | CdH2O2 | 146.43 g/mol

Supplier:  Thermo Scientific Chemicals 01066209

 View more versions of this product

Catalog No. AA1066209


Only null left
Explore available promotions

Description

Description

It is found in the industrial nickel cadmium storage batteries, in cadmium plating, in the manufacture of cadmium salts, laboratory reagents. It is a useful biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cadmium hydroxide
21041-95-2
10 g
CdH2O2
UN2570
Soluble in dilute acids. Slightly soluble in water; 0.026g/100 mL.
[OH-].[OH-].[Cd++]
146.43
99.98%
Air Sensitive
300°C (decomposition)
Powder
(metals basis)
MFCD00015995
14,1620
PLLZRTNVEXYBNA-UHFFFAOYSA-L
cadmium(2+) dihydroxide
146.41
Puratronic
4.79 g/mL at 15°C

Chemical Identifiers

21041-95-2
146.43
PLLZRTNVEXYBNA-UHFFFAOYSA-L
[OH-].[OH-].[Cd++]
CdH2O2
MFCD00015995
cadmium(2+) dihydroxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c

H330-H341-H350-H361fd-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS

EINECSNumber : 244-168-5

RTECSNumber : EV1260000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only