accessibility menu, dialog, popup

UserName

CAS RN 21041-95-2

IUPAC Name: cadmium(2+) dihydroxide
Synonyms: cadmium hydroxide
Molecular Weight (g/mol): 146.43
Molecular Formula: CdH2O2
InChi Key: PLLZRTNVEXYBNA-UHFFFAOYSA-L
SMILES: [OH-].[OH-].[Cd++]
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results