missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Granular, For Analysis, Particle size about 3-6mm, EMSURE™, MilliporeSigma™

Catalog No. M1020040250
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1020040250 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1020040250 Supplier Technidata Supplier No. 1.02004.0250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]

Specifications

Appearance Metallic solid
Quantity 250 g
Odor Odorless
Packaging Metal can
Form Solid
UN Number UN 3288
Formula Weight 112.41
Chemical Name or Material Cadmium
Hazard Category H350: May cause cancer. H330: Fatal if inhaled. H341: Suspected of causing genetic defects. H361fd: Suspected of damaging fertility. Suspected of damaging the unborn child.
H372: Causes damage to organs through prolonged or repeated exposure.
H410: Very toxic to aquatic life with long lasting effects.