Learn More
Cadmium foil, 1.0mm (0.04 in.) thick, 15x15cm (5.9x5.9 in.), 99.95% (metals basis), Thermo Scientific Chemicals
$1334.58 - $1334.58
Chemical Identifiers
| CAS | 7440-43-9 |
|---|---|
| Molecular Formula | Cd |
| Molecular Weight (g/mol) | 112.41 |
| MDL Number | MFCD00010914 |
| InChI Key | BDOSMKKIYDKNTQ-UHFFFAOYSA-N |
| Synonym | kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds |
| PubChem CID | 23973 |
| ChEBI | CHEBI:22977 |
| IUPAC Name | cadmium |
| SMILES | [Cd] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-43-9 | |
| 112.41 | |
| BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
| 23973 | |
| cadmium |
| Cd | |
| MFCD00010914 | |
| kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds | |
| CHEBI:22977 | |
| [Cd] |
Specifications
| 7440-43-9 | |
| ≈195g/15x15cm | |
| Cd | |
| kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds | |
| [Cd] | |
| 112.41 | |
| CHEBI:22977 | |
| Cadmium foil, 1.0mm (0.04 in.) thick, 15x15cm (5.9x5.9 in.) |
| 1 Pc. | |
| Foil | |
| MFCD00010914 | |
| BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
| cadmium | |
| 23973 | |
| 99.95% | |
| (metals basis) |
Safety and Handling
GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c
H330-H341-H350-H361fd-H372-H500
EINECSNumber : 231-152-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only