missing translation for 'onlineSavingsMsg'
Learn More

Cadmium foil, 0.25mm (0.01in) thick, Puratronic™, 99.9975% (metals basis)

Catalog No. p-7048734
Change view
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11370FI 50 x 50 mm
AA11370GH 100 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA11370FI Supplier Thermo Scientific Chemicals Supplier No. 011370FI
Only null left

CAS: 7440-43-9 | Cd | 112.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
SMILES [Cd]

Specifications

Quantity 50 x 50 mm
Weight ≈5.4g/50x50mm
Form Foil
IUPAC Name cadmium
Percent Purity 99.9975%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Cadmium foil, 0.25mm (0.01 in.) thick
Hazard Category H330-H341-H350-H361fd-H372-H500
Hazard Statement GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c
EINECSNumber 231-152-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only