Learn More
Cadmium fluoride, Puratronic™, 99.99% (metals basis)
CAS: 7790-79-6 | CdF2 | 150.41 g/mol
$850.88 - $850.88
Chemical Identifiers
| CAS | 7790-79-6 |
|---|---|
| Molecular Formula | CdF2 |
| Molecular Weight (g/mol) | 150.41 |
| MDL Number | MFCD00010919 |
| InChI Key | LVEULQCPJDDSLD-UHFFFAOYSA-L |
| IUPAC Name | cadmium(2+) difluoride |
| SMILES | [F-].[F-].[Cd++] |
Description
Cadmium fluoride is used in phosphors, glass and nuclear reactor controls. It is also used in high-temperature lubricants and to start crystals for lasers. It is used in oxygen-sensitive applications such as the production of metallic alloys and it is also used in limited medical treatment protocols. In addition, it can be used in synthetic organic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7790-79-6 | |
| 150.41 | |
| LVEULQCPJDDSLD-UHFFFAOYSA-L | |
| [F-].[F-].[Cd++] |
| CdF2 | |
| MFCD00010919 | |
| cadmium(2+) difluoride |
Specifications
| 1758°C | |
| 7790-79-6 | |
| 100 g | |
| MFCD00010919 | |
| 14,1619 | |
| LVEULQCPJDDSLD-UHFFFAOYSA-L | |
| cadmium(2+) difluoride | |
| 150.4 | |
| Puratronic | |
| 6.64 g/mL | |
| (metals basis) |
| 1100°C | |
| Powder | |
| CdF2 | |
| UN2570 | |
| Soluble in water (43.5g/L). | |
| [F-].[F-].[Cd++] | |
| 150.41 | |
| 99.99% | |
| Odorless | |
| Cadmium fluoride |
Safety and Handling
GHS H Statement
H301-H330-H340-H350-H360-H372
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
H301-H330-H340-H350-H360FD-H372
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 232-222-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only