missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride, ultra dry, 99.998% (metals basis)

Catalog No. p-7049275
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1366706 5 g
AA1366714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1366706 Supplier Thermo Scientific Chemicals Supplier No. 01366706
Only null left

CAS: 10108-64-2 | CdCl2 | 183.31 g/mol

Cadmium chloride is utilized in the preparation of CdTe nanocrystals, highly luminescent. It is also employed in analytical chemistry, photography, dyeing and calico printing, ingredient of electroplating baths, manufacture of special mirrors, the preparation of cadmium standard, analysis of sulfides, and testing pyridine bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon

Chemical Identifiers

CAS 10108-64-2
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00010916
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
PubChem CID 24947
ChEBI CHEBI:35456
IUPAC Name dichlorocadmium
SMILES [Cl-].[Cl-].[Cd++]

Specifications

Boiling Point 960°C
Melting Point 568°C
Physical Form Beads
Quantity 5 g
UN Number UN2570
Merck Index 14,1617
Solubility Information Soluble in water and acetone.
Formula Weight 183.32
Percent Purity 99.998%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.047 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium chloride, Ultra dry
Show More Show Less
Hazard Category H301-H330-H335-H340-H350-H360FD-H372
Hazard Statement GHS H Statement
H301-H330-H340-H350-H360-H372
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber 233-296-7
RTECSNumber EV0175000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only