Learn More
Cadmium chloride, ACS reagent, anhydrous
CAS: 10108-64-2 | CdCl2 | 183.31 g/mol
Supplier: Thermo Scientific Chemicals 315271000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 960.0°C | |
| 568.0°C | |
| 10108-64-2 | |
| Powder | |
| CdCl2 | |
| MFCD00010916 | |
| 15,1622 | |
| Solubility in water: 1400g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in methanol and ethanol, practically insoluble in ether | |
| [Cl-].[Cl-].[Cd++] | |
| 183.31 | |
| CHEBI:35456 | |
| ≥99.0% | |
| Plastic bottle |
| Cadmium chloride | |
| Anhydrous, For analysis ACS | |
| White | |
| 100 g | |
| CdCl2 | |
| 04,485 | |
| cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride | |
| YKYOUMDCQGMQQO-UHFFFAOYSA-L | |
| dichlorocadmium | |
| 24947 | |
| 183.31 | |
| ACS Reagent |
Chemical Identifiers
| 10108-64-2 | |
| 183.31 | |
| YKYOUMDCQGMQQO-UHFFFAOYSA-L | |
| 24947 | |
| dichlorocadmium |
| CdCl2 | |
| MFCD00010916 | |
| cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride | |
| CHEBI:35456 | |
| [Cl-].[Cl-].[Cd++] |
Safety and Handling
GHS H Statement
May cause genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
May cause cancer.
May damage fertility.
May damage the unborn child.
Toxic if swallowed
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF S
GHS Signal Word: Danger
EINECSNumber : 233-296-7
RUO – Research Use Only