missing translation for 'onlineSavingsMsg'
Learn More

Cadmium carbonate, Puratronic™, 99.998% (metals basis)

Catalog No. p-4781825
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1066009 10 g
AA1066018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1066009 Supplier Thermo Scientific Chemicals Supplier No. 01066009
Only null left

CAS: 513-78-0 | CCdO3 | 172.42 g/mol

Cadmium carbonate is the raw material for the production of pigments for ceramic and frits and for production of cadmium salt. Used as reagents. It is used for intermediates of manufacturing polyester, glass pigment flux, organic catalysts, plastic plasticizer, stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 513-78-0
Molecular Formula CCdO3
Molecular Weight (g/mol) 172.42
MDL Number MFCD00010918
InChI Key GKDXQAKPHKQZSC-UHFFFAOYSA-L
Synonym cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
PubChem CID 10564
IUPAC Name cadmium(2+);carbonate
SMILES [Cd++].[O-]C([O-])=O

Specifications

Melting Point 500°C (decomposition)
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.998% (Metals basis)
Solubility Information Soluble in dilute acids,concentrated solutions of ammonium salts. Insoluble in water.
Formula Weight 172.42
Odor Odorless
Density 4.26 g/cm3
Chemical Name or Material Cadmium carbonate
Hazard Category H302+H312-H330-H340-H350-H361-H372
Hazard Statement GHS H Statement
H302-H312-H332
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P312-P302+P352-P304+P340-P308+P313-P310-P312-P330-P363-P501c
DOTInformation DOT Class: 6.1; Packaging Group: II
EINECSNumber 208-168-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only