missing translation for 'onlineSavingsMsg'
Learn More

Cadmium carbonate, >99% (metals basis), Thermo Scientific Chemicals

$201.25 - $201.25

Chemical Identifiers

CAS 513-78-0
Molecular Formula CCdO3
Molecular Weight (g/mol) 172.42
MDL Number MFCD00010918
InChI Key GKDXQAKPHKQZSC-UHFFFAOYSA-L
Synonym cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
PubChem CID 10564
IUPAC Name cadmium(2+);carbonate
SMILES [Cd++].[O-]C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1222930
View Documents
Thermo Scientific Chemicals
01222930
250 g
Each for $201.25
Only null left
 
Description

Description

Cadmium carbonate is the raw material for the production of pigments for ceramic and frits. Used as reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

513-78-0
172.42
GKDXQAKPHKQZSC-UHFFFAOYSA-L
10564
[Cd++].[O-]C([O-])=O
CCdO3
MFCD00010918
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
cadmium(2+);carbonate

Specifications

500°C (decomposition)
White
250 g
CCdO3
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
GKDXQAKPHKQZSC-UHFFFAOYSA-L
cadmium(2+);carbonate
10564
Odorless
4.26 g/cm3
513-78-0
Powder
>99% (Metal basis)
MFCD00010918
Very slightly soluble in water. Soluble in dilute acids,concentrated solutions of ammonium salts.
[Cd++].[O-]C([O-])=O
172.42
172.42
White powder
Cadmium carbonate
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P312-P302+P352-P304+P340-P308+P313-P310-P312-P330-P363-P501c

H302+H312-H330-H340-H350-H361-H372

DOTInformation : DOT Class: 6.1; Packaging Group: II

EINECSNumber : 208-168-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only