missing translation for 'onlineSavingsMsg'
Learn More

Cadmium carbonate, 97%

Catalog No. AAA1581218
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1581218 50 g
AAA1581230 250 g
AAA158120B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1581218 Supplier Thermo Scientific Chemicals Supplier No. A1581218
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 513-78-0 | CCdO3 | 172.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 513-78-0
Molecular Formula CCdO3
Molecular Weight (g/mol) 172.42
MDL Number MFCD00010918
InChI Key GKDXQAKPHKQZSC-UHFFFAOYSA-L
Synonym cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
PubChem CID 10564
SMILES [Cd++].[O-]C([O-])=O

Specifications

Melting Point 500°C (decomposition)
Quantity 50 g
UN Number UN2570
Merck Index 14,1616
IUPAC Name cadmium(2+) carbonate
Formula Weight 172.42
Percent Purity 97%
Odor Odorless
Density 4.26
Chemical Name or Material Cadmium carbonate
Hazard Category H302+H312-H330-H340-H350-H361-H372
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P312-P302+P352-P304+P340-P308+P313-P310-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 208-168-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only