missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Acetate Dihydrate, Reagent, Reagents

Catalog No. 01338068
Change view
Click to view available options
Quantity:
25 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338068 25 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01338068 Supplier Labchem Inc Supplier No. C115725025LB

missing translation for 'validMainframeMsg'

Cadmium Acetate Dihydrate, Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H10CdO6
Molecular Weight (g/mol) 266.53
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L
IUPAC Name cadmium(2+) diacetate dihydrate
SMILES O.O.[Cd++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 256°C
Color Colorless/White
pH 7.1
Physical Form Solid
Quantity 25 lb.
Grade Reagent
Packaging Poly Pail
Specific Gravity 2.34
Chemical Name or Material Cadmium Acetate Dihydrate, Reagent