Learn More
Cadmium acetate dihydrate, 99.999% (metals basis)
CAS: 5743-04-4 | C4H10CdO6 | 266.53 g/mol
$674.34 - $674.34
Chemical Identifiers
| CAS | 5743-04-4 |
|---|---|
| Molecular Formula | C4H10CdO6 |
| Molecular Weight (g/mol) | 266.53 |
| MDL Number | MFCD00012449 |
| InChI Key | AUIZLSZEDUYGDE-UHFFFAOYSA-L |
Description
It is used in dyestuff intermediates, ceramics and producing iridescent effects on porcelains and pottery. It acts as a reagent for determination of S, Se, and Te. It is also used in cadmium electroplating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| White | |
| Powder | |
| 99.999% (Metals basis) | |
| MFCD00012449 | |
| AUIZLSZEDUYGDE-UHFFFAOYSA-L | |
| 266.54 (230.50 Anhydrous) | |
| Hygroscopic | |
| Cadmium acetate dihydrate |
| 5743-04-4 | |
| 100 g | |
| C4H10CdO6 | |
| Soluble in water,ethanol and methanol. | |
| 266.53 | |
| Odorless | |
| 2.01 |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H335-H350-H373
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 208-853-2
RTECSNumber : AF7505000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only