Learn More
Cadmium acetate dihydrate, 98%
In cadium electroplating | CAS: 5743-04-4 | C4H10CdO6 | 266.53 g/mol
$117.04 - $1023.88
Chemical Identifiers
| CAS | 5743-04-4 |
|---|---|
| Molecular Formula | C4H10CdO6 |
| Molecular Weight (g/mol) | 266.53 |
| MDL Number | MFCD00012449 |
| InChI Key | AUIZLSZEDUYGDE-UHFFFAOYSA-L |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4158122
|
Thermo Scientific Chemicals
04158122 |
100 g |
Each for $117.04
|
|
|||||
|
AA4158136
|
Thermo Scientific Chemicals
04158136 |
500 g |
Each for $317.75
|
|
|||||
|
AA41581A3
|
Thermo Scientific Chemicals
041581A3 |
2 kg |
Each for $1,023.88
|
|
|||||
Description
Cadmium acetate dihydrate is in the porcelain production for the production of iridescent used effects. Moreover, it is in the analytical chemistry, a reagent for the detection of sulfur, selenium and tellurium. It is also used as chemical and pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| White | |
| Powder | |
| C4H10CdO6 | |
| UN2570 | |
| Freely soluble in water, soluble in alcohol | |
| 266.53 | |
| 98% | |
| 2.01 g/mL |
| 5743-04-4 | |
| 100 g | |
| MFCD00012449 | |
| 14,1614 | |
| AUIZLSZEDUYGDE-UHFFFAOYSA-L | |
| 266.54 (230.50 Anhydrous) | |
| Hygroscopic | |
| Cadmium acetate dihydrate |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H335-H350-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber : 208-853-2
RTECSNumber : AF7505000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only